Learn More
Dutogliptin, MedChemExpress
Product Code. 16483758
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
Product Code. 16483758
Brand: Medchem Express HY10286
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Dutogliptin (PHX-1149 free base) is an orally available, potent, and selective dipeptidyl peptidase-4 (DPP4) inhibitor for the treatment of type 2 diabetes mellitus.
Dutogliptin (PHX-1149 free base) is an orally available, potent, and selective dipeptidyl peptidase-4 (DPP4) inhibitor for the treatment of type 2 diabetes mellitus.
Specifications
Chemical Name or Material Dutogliptin
Physical Form Solid
CAS 852329-66-9
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C10H20BN3O3
For Use With (Application) Metabolism-protein/nucleotide metabolism
Synonym PHX-1149 free base
Solubility Information DMSO : 300 mg/mL (1244.30 mM; Need ultrasonic)
SMILES O=C(CN[C@@H]1CCNC1)N(CCC2)[C@@H]2B(O)O
Molecular Weight (g/mol) 241.1
Formula Weight 241.1
Percent Purity 99.16%
Grade Research