missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ DUSP7 (Human) Recombinant Protein
Human DUSP7 full-length ORF ( AAH19107.1, 1 a.a. - 320 a.a.) recombinant protein with GST-tag at N-terminal.
Marke: Abnova™ H00001849-P01.25ug
Weitere Details : Gewicht : 0.00010kg
Beschreibung
- Sequence: MLRRLRKGNLPIRSIIPNHADKERFATRCKAATVLLYDEATAEWQPEPGAPASVLGLLLQKLRDDGCQAYYLQGGFNKFQTEYSEHCETNVDSSSSPSSSPPTSVLGLGGLRISSDCSDGESDRELPSSATESDGSPVPSSQPAFPVQILPYLYLGCAKDSTNLDVLGKYGIKYILNVTPNLPNAFEHGGEFTYKQIPISDHWSQNLSQFFPEAISFIDEARSKKCGVLVHCLAGISRSVTVTVAYLMQKMNLSLNDAYDFVKRKKSNISPNFNFMGQLLDFERTLGLSSPCDNHASSEQLYFSTPTNHNLFPLNTLEST
Spezifikation
AAH19107.1 | |
dual specificity phosphatase 7 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DUSP7 | |
GST |
1849 | |
Wheat germ expression system | |
25 μg | |
MKP-X, MKPX, PYST2 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |