missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DUSP19 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA550222
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total DUSP19 protein.
Dual-specificity phosphatases.Specifications
DUSP19 | |
Polyclonal | |
Unconjugated | |
DUSP19 | |
5930436K22Rik; C79103; dual specificity phosphatase 19; dual specificity phosphatase TS-DSP1; Dual specificity protein phosphatase 19; DUSP17; DUSP19; LMWDSP3; LMW-DSP3; low molecular weight dual specificity phosphatase 3; Protein phosphatase SKRP1; SAPK pathway-regulating phosphatase 1; SKRP1; Stress-activated protein kinase pathway-regulating phosphatase 1; TS-DSP1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
142679, 68082 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1.2 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q8K4T5, Q8WTR2 | |
DUSP19 | |
Full length fusion protein of Human DUSP19 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |