missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DUS1L Polyclonal Antibody
GREENER_CHOICE

Product Code. 15752711 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15752711 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15752711 Supplier Invitrogen™ Supplier No. PA554783

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: LTVHGRTKEQ KGPLSGAASW EHIKAVRKAV AIPVFANGNI QCLQDVERCL RDTGVQGVMS AEGNLHNPAL FEGRSPAVWE LAEEYLDIVR EH Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 90%.

Catalyzes the synthesis of dihydrouridine, a modified base found in the D-loop of most tRNAs. Target protein has a region of similarity to tRNA dihydrouridine synthase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DUS1L
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Dus1l
Gene Accession No. Q6P1R4
Gene Alias 1110032N12Rik; dihydrouridine synthase 1 like; dihydrouridine synthase 1-like; dihydrouridine synthase 1-like (S. cerevisiae); DUS1; DUS1L; embryo-related protein; liver regeneration-related protein LRRG08/LRRG09; Pp3111; tRNA-dihydrouridine synthase 1-like; tRNA-dihydrouridine(16/17) synthase [NAD(P)(+)]-like; x85
Gene Symbols Dus1l
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DUS1L. Recombinant protein control fragment (Product #RP-93410).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64118
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.