Learn More
DWK Life Sciences DURAN™ Woulff Bottle, 3 standard ground necks
Product Code. 10510072 Shop All DWK Life Sciences Products
Click to view available options
Capacity (Metric):
500 mL
1000 mL
2000 mL
5000 mL
Unit Size:
Each
Product Code. 10510072
Brand: DWK Life Sciences 247097306
Only null left
Add to basket
Add to basket
The Woulff bottle is used in vacuum apparatus and serves to collect, clean and/or dry gases.
Woulff''s bottles are part of vacuum apparatus, in which they are used on the one hand as collecting vessels for gas generation or for cleaning and/or drying gases. As part of a vacuum apparatus, Woulff''s bottles are suitable for vacuum applications and often also used as an intermediate vessel to prevent spontaneous drop-off of the vacuum and the associated liquid suction back into the vacuum pump.
  • Tight closure due to standard ground joint
  • Vacuum resistant due to the wall thickness and geometry
  • Three standard ground joints for the import or export of gases
  • Optimum safety for working under pressure or vacuum
  • Fired-on, highly durable white print
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 5000 mL
For Use With (Application) Collecting and dissolving gases in liquids; Production of gases; Safe working under vacuum; In series: Purification of a gas
Material Borosilicate Glass (3.3)
No. per Pack 1/Pk.
Diameter (Metric) 185 mm
Autoclavable Autoclavable