Learn More
DWK Life Sciences DURAN™ Tube with Screw Thread, with DIN thread
Shop All DWK Life Sciences Products
Click to view available options
Thread Size:
GL 14
GL 18
GL 25
GL 32
GL 45
missing translation for 'unitSize'
Each
Pack of 10
Description
This threaded tube has a screw thread and can be incorporated into glass components by a further processing step.
- With DIN thread
Specifications
Specifications
| Material | Borosilicate Glass (3.3) |
| For Use With | Shoring in glass apparatuses |
| Quantity | 1 Ea. |
| Thread Size | GL 45 |
| Autoclavable | Autoclavable |
| Diameter (Metric) | 40 mm |
| Wall Thickness | 2.3 mm |