missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences DURAN™ Steriplan Petri Dishes
Shop All DWK Life Sciences Products

Click to view available options
Diameter (Metric):
100 mm
200 mm
Height (Metric):
10 mm
20 mm
30 mm
Unit Size:
Each
Description
- Soda-lime glass made
- Autoclavable
- For cultivation of microorganism
Specifications
Specifications
| Material | Soda Lime Glass |
| Height (Metric) | 20 mm |
| Diameter (Metric) | 100 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction