Learn More
DWK Life Sciences DURAN™ Plain Cylinder, with round base, without graduation
Shop All DWK Life Sciences Products
Click to view available options
Capacity (Metric):
80 mL
190 mL
440 mL
550 mL
Unit Size:
Pack of 10
Description
The multitude of different work steps in the laboratory environment demands a multitude of universally applicable glassware. These glass devices usually have a very simple shape and are characterized by their versatility.
- Plain ground rim
- With round base
Specifications
Specifications
| Material | Borosilicate Glass (3.3) |
| Diameter (Metric) Outside | 60 mm |
| Capacity (Metric) | 440 mL |
| For Use With (Application) | Diluting solutions; Mixing several components |
| Includes | 10/Pk. |
| Autoclavable | Autoclavable |