Learn More
DWK Life Sciences DURAN™ KPG™ Stirrer Shaft, interchangeable
Product Code. p-7189253 Shop All DWK Life Sciences Products
Click to view available options
Type:
Ground Joint
Polished
Unit Size:
Pack of 50
Product Code. 15884638
Brand: DWK Life Sciences 286305701
Only null left
Add to basket
Add to basket
Use as an alternative to magnetic stirrers with high viscosity or high stirring speeds.
Solid DURAN™ KPG™ stirrer shafts are used for further processing into a wide variety of stirrers. They are available in the qualities ground and polished. The bearing sleeve and agitator shaft are precisely matched to each other by precision grinding.
  • Solid
  • Available in polished and ground version
  • The bearing sleeve and agitator shaft are precisely matched to each other by precision grinding
TRUSTED_SUSTAINABILITY
Specifications
Quantity 50/Pk.
Item Description Stirring of reaction solutions with high viscosities or at high stirring speeds; For further processing to various stirrer shapes and types
Type Ground Joint