Learn More
DWK Life Sciences DURAN™ Glass Stopper, Clear, Semi-hollow, Standard Taper Ground Joint, Octagonal flat head
Clear DURAN™ glass ground stopper.
21.45 € - 62.75 €
Specifications
Autoclavable | No |
---|---|
Material | Borosilicate Glass |
For Use With (Application) | Storage of air sensitive reagents, and chemicals; Storage of hygroscopic solids, granules and powders; Storage of pharmaceutical APIs; Sealing laboratory glassware such as flasks, reagent bottles, and mixing cylinders |
Quantity | 1 Ea. |
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | |||||
10263641
|
DWK Life Sciences
216241106 |
ST 34/35 |
21.45 €
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
10010141
|
DWK Life Sciences
216241603 |
ST 85/55 |
62.75 €
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
10315351
|
DWK Life Sciences
216241209 |
ST 45/40 |
22.35 €
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
15632264
|
DWK Life Sciences
216241303 |
ST 60/46 |
31.45 €
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
Semi-hollow or solid DURAN™ glass stoppers can be used to close glassware items such as flasks, separatory funnels or reagent bottles with ground joint necks. The Standard Taper ground glass joint forms a tight, leak-proof seal. Use grease for heating or vacuum applications. With octagonal shaped head. Note on using DURAN™ glass stoppers with soda-lime glass bottles: If the bottle and the stopper have a temperature difference greater than 30°C, the glass stoppers can become stuck!
- All DURAN™ glass components and therefore suitable for storage of aggressive materials
- Tight closure due to Standard Taper ground joint
- Octagonal shaped head
Specifications
No | |
Borosilicate Glass | |
Storage of air sensitive reagents, and chemicals; Storage of hygroscopic solids, granules and powders; Storage of pharmaceutical APIs; Sealing laboratory glassware such as flasks, reagent bottles, and mixing cylinders | |
1 Ea. |