Learn More
Lenz Laborglasinstrumente™ Duran Glass Bent Gas Inlet Tubes
Product Code. 10250881
Change view
Click to view available options
Length (Metric):
250mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10250881 250mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10250881 Supplier Lenz Laborglasinstrumente™ Supplier No. 05083001
  • ONLINE EXCLUSIVE PRICE
    24.00 €
    13.32 € / Pack of 1
    Save 10.68 €
    44% Off
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Includes NS cone and thin capillary
Length (Metric) 250mm
Type Gas Inlet Tube
Material Duran Glass
Quantity 1Each