Learn More
DWK Life Sciences DURAN™ Filter Apparatus, Witt Type, complete, with interchangeable lid, and KECK™ assembly set, standard ground joint 45/40
The Witt filtration apparatus is used for solid-liquid filtration in which the filtrate is subsequently further processed.
371.00 € - 469.00 €
Specifications
Autoclavable | Autoclavable |
---|---|
Material | Borosilicate Glass (3.3), PBT, Polypropylene, VMQ, EPDM |
For Use With (Application) | Filtering; Preferably collecting the filtrate; Filtration at reduced pressure; Vacuum filtration; Safe working under vacuum |
Quantity | 1 Each |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
10414081
|
DWK Life Sciences
247315703 |
NS 45/40, DN 150 |
469.00 €
Each |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
10570012
|
DWK Life Sciences
247314604 |
NS 45/40, DN 100 |
371.00 €
Each |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
Description
One of the most important separation methods in the laboratory environment is filtration, which is used to separate or clean different media. For this working method, filters are required which are selected according to the application with regard to size and porosity. Buechner funnels are cylinders with an integrated large-pored sieve plate. Without additional filter paper, only coarse filtration is possible. Buechner funnels can also be used in vacuum filtration. The applied vacuum accelerates the filtration.
- Tight closure due to standard ground joint
- Wide rough-ground tubulature
- Fired-on, highly durable white print
Specifications
Autoclavable | |
Borosilicate Glass (3.3), PBT, Polypropylene, VMQ, EPDM | |
Filtering; Preferably collecting the filtrate; Filtration at reduced pressure; Vacuum filtration; Safe working under vacuum | |
1 Each |