Learn More
Hirschmann™ DURAN™ Burette Tubes with Blue Graduations
Description
- Short line
- Blue graduation
Specifications
Specifications
| Diameter (Metric) Outer | 8 mm |
| Capacity (Metric) | 50 mL |
| Length (Metric) | 500 mm |
| For Use With (Equipment) | Buret Stopcock |
| Material | Borosilicate Glass |
| No. per Pack | 2/Pack |
| Scale | Blue |
| Tolerance | ±0.075 mL |
| Type | Buret Tube |