Learn More
Hirschmann™ DURAN™ Burette Tubes with Blue Graduations
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
Unit Size:
Pack of 2
Description
- Short line
- Blue graduation
Specifications
Specifications
| Color | Blue |
| Diameter (Metric) Outer | 8 mm |
| Capacity (Metric) | 25 mL |
| Length (Metric) | 500 mm |
| For Use With (Equipment) | Burette Stopcock |
| Material | Borosilicate Glass |
| No. per Pack | 2/Pack |
| Scale | Blue |
| Type | Buret Tube |