Learn More
Hirschmann™ DURAN™ Burette Tubes with Blue Graduations
Product Code. 13177959
Change view
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
Unit Size:
Pack of 2
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
13177959 10 mL Pack of 2
13187959 25 mL Pack of 2
13197959 50 mL Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 13177959 Supplier Hirschmann™ Supplier No. 3071260
Only null left
Add to basket
Add to basket
Burette assembly with burette tube and separate PTFE stopcock with PP tip. Excellent for titrating exact amounts of chemical solutions in the laboratory.
  • Short line
  • Blue graduation
Specifications
Diameter (Metric) Outer 8 mm
Capacity (Metric) 10 mL
Length (Metric) 500 mm
For Use With (Equipment) Buret Stopcock
Material Borosilicate Glass
No. per Pack 2/Pack
Scale Blue
Tolerance ±0.03 mL
Type Buret Tube