Learn More
Hirschmann™ DURAN™ Burette Stopcock Straight
Product Code. 10547451
Change view
Click to view available options
For Use With (Equipment):
Volume of burette 2 - 10 mL
Volume of burette 25 mL
Volume of burette 50 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
10547451 Volume of burette 25 mL Each
10489132 Volume of burette 2 - 10 mL Each
10709391 Volume of burette 50 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10547451 Supplier Hirschmann™ Supplier No. 8900870
Only null left
Add to basket
Add to basket
Spare part for the economical repair of a burette with straight stopcock.
  • With glass key
Specifications
Type Straight Burette Stopcock
Description With glass key
Material Duran Glass
No. per Pack 1 Each
For Use With (Equipment) Volume of burette 25 mL