Learn More
DWK Life Sciences™ DURAN™ Blank for Round Bottom Cylinder, for jacketed vessel, rim cracked-off, without print
Product Code. 10385581 Shop All DWK Life Sciences Products
Click to view available options
Diameter (Metric) Outer:
105 mm
130 mm
160 mm
185 mm
215 mm
250 mm
Unit Size:
Each
Product Code. 10385581
Brand: DWK Life Sciences™ 210276678
Nur noch null auf Lager
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
For processing as a reaction vessel.
DURAN™ cylinders with a round bottom are convincing due to their good wall thickness distribution and enable the cost-effective production of reaction vessels with reproducible geometry. For pressure resistance, please refer to our notes in the technical appendix of the laboratory glassware catalogue. For glass articles which deviate from the original form due to further processing, the permissible operating data must be determined in accordance with the technical rules and regulations.
  • For jacket vessels edge blown off
  • Component for further processing into reaction vessels
  • Cost-effective production of reaction vessels with reproducible geometry
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Spezifikation
Description Autoclavable
Diameter (Metric) Outer 250 mm
For Use With (Application) For processing as a reaction vessel
Includes 1/Pk.
Depth (Metric) Interior 5.5 mm wall thickness
Material Borosilicate Glass (3.3)