Learn More
DUPA(OtBu)-OH, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
DUPA(OtBu)-OH is a DUPA precursor. DUPA is used as the targeting moiety to actively deliver Docetaxel (DTX) for treatment of Prostate-Specific Membrane Antigen (PSMA) expressing prostate cancer.
Specifications
Specifications
| Chemical Name or Material | DUPA(OtBu)-OH |
| Physical Form | Solid |
| CAS | 1026987-94-9 |
| Color | White |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C23H40N2O9 |
| For Use With (Application) | Cancer-programmed cell death |
| Solubility Information | DMSO : 125 mg/mL (255.85 mM; Need ultrasonic) |
| SMILES | O=C(OC(C)(C)C)CC[C@@H](C(OC(C)(C)C)=O)NC(N[C@H](C(OC(C)(C)C)=O)CCC(O)=O)=O |
| Show More |