missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Duligotuzumab Recombinant Monoclonal Antibody
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Duligotuzumab has been used in trials studying the treatment of Neoplasms, Colorectal Cancer, Head and Neck Cancer, and Epithelial Tumors, Malignant.
Specifications
Specifications
| Antigen | Duligotuzumab |
| Applications | ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance |
| Classification | Recombinant Monoclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 |
| Gene Alias | MEHD7945A; RG-7597 |
| Host Species | Human |
| Purification Method | Protein A |
| Quantity | 1 mg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?