Learn More
Fisherbrand™ Anodized Dual Aluminum Blocks
Product Code. 11452107
Change view
Click to view available options
Product Type:
D1196, 96-Well
D12384, 384 WELL PCR PLATE
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type unitSize
11452107 D12384, 384 WELL PCR PLATE Each
11401997 D1196, 96-Well Each
11411997 D1196, 96-Well Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11452107 Supplier Fisherbrand™ Supplier No. D12384
Please to purchase this item. Need a web account? Register with us today!
Useful for a variety of applications in molecular biology, histology, clinical, environmental and industrial. Fisherbrand™ Anodized Dual Aluminum Blocks are for use with Fisherbrand Digital Block Heaters.
  • Excellent heat transfer
  • Anodized to resist thermal shock
  • For holding test tubes for digital heaters
  • Available to fit a range of tube counts and sizes
Especificaciones
For Use With (Equipment) PCR
Product Type D12384, 384 WELL PCR PLATE
Quantity 1 CT
Weight 1 kg
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.