missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
DTX1/DTX4 Monoclonal antibody specifically detects DTX1/DTX4 in Human samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF
Specifications
Specifications
| Antigen | DTX1/DTX4 |
| Applications | Western Blot, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF |
| Classification | Monoclonal |
| Clone | 730612 |
| Conjugate | FITC |
| Dilution | Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready |
| Formulation | PBS |
| Gene Alias | DTX1;Fxit1;hDx-1;mKIAA4160 |
| Host Species | Mouse |
| Immunogen | E. coli-derived recombinant human DTX1/DTX4, Met1-Phe147, Accession # Q86Y01 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?