missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dtk/TYRO3 Antibody (96201) [DyLight 405], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB859E
This item is not returnable.
View return policy
Description
Dtk/TYRO3 Monoclonal antibody specifically detects Dtk/TYRO3 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Gene Knock-Out
Specifications
| Dtk/TYRO3 | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Gene Knock-Out | |
| 96201 | |
| Flow Cytometry, Immunohistochemistry, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Knockout Validated | |
| Brt, BYK, DTK, EC 2.7.10, EC 2.7.10.1, FLJ16467, RSE, Sky, Tif, TYRO3 protein tyrosine kinase, Tyrosine-protein kinase byk, Tyrosine-protein kinase DTK, tyrosine-protein kinase receptor TYRO3, Tyrosine-protein kinase RSE, Tyrosine-protein kinase SKY | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Tyro3/Dtk, Ala41-Ser428 (predicted), Accession # Q06418 | |
| 0.1 mL | |
| Cancer, Neuroscience, Neurotransmission, Protein Kinase, Tyrosine Kinases | |
| 7301 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction