missing translation for 'onlineSavingsMsg'
Learn More

DSG4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Product Code. 16882185 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16882185 20 μL 20µL
16872185 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16882185 Supplier Proteintech Supplier No. 230871AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Desmoglein proteins are cadherin-type cell adhesion molecules. Desmoglein 4 (dsg4) shares 41% identity with human desmoglein 1, 37% with human desmoglein 2 and 50% with human desmoglein 3. A type I membrane protein of the cadherin protein family, dsg4 is expressed in salivary gland, suprabasal epidermis, hair follicle, testis, prostate and skin. In the hair follicle, dsg4 is an important mediator of keratinocyte cell adhesion and coordinates the transition from proliferation to differentiation. The human DSG4 gene is composed of 16 exons spanning approximately 37 kb of 18q12 and is situated between DSG1 and DSG3.

Specifications

Antigen DSG4
Applications Western Blot
Classification Polyclonal
Concentration 0.63 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DSG4
Gene Accession No. Q7TMD7, Q86SJ6
Gene Alias Cadherin family member 13, CDGF13, CDHF13, desmoglein 4, DSG4, LAH
Gene Symbols DSG4
Host Species Rabbit
Immunogen DSG4 Fusion Protein Ag19520
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 147409, 16769
Target Species Mouse, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.