Learn More
Miele Drying unit, electro for 900 mm wide appliances and optimum interior / exterior drying of the dishes
Brand:  Miele 12678660
Drying unit, electro for 900 mm wide appliances and optimum interior / exterior drying of the dishes
Product Code. 16302248
  • ONLINE EXCLUSIVE PRICE
    9715.00 €
    9589.68 € / Each
    Save 125.32 €
    1% Off
Only null left
Estimated Shipment: 29-01-2026