Learn More
Drotaverine Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | Drotaverine Hydrochloride |
| CAS | 985-12-6 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C24 H31 N O4 . Cl H |
| Quantity | 10 mg |
| Synonym | Isoquinoline, 1-[(3,4-diethoxyphenyl)methylene]-6,7-diethoxy-1,2,3,4-tetrahydro-, hydrochloride (1:1), Isoquinoline, 1-(3,4-diethoxybenzylidene)-6,7-diethoxy-1,2,3,4-tetrahydro-, hydrochloride (7CI,8CI), Isoquinoline, 1-[(3,4-diethoxyphenyl)methylene]-6,7-diethoxy-1,2,3,4-tetrahydro-, hydrochloride (9CI), Drotaverin hydrochloride, Drotaverine hydrochloride, Drotaverinium chloride, Drotin, Isodihydroperparine hydrochloride, NOSH-BRA, No-Spa, No-Spa hydrochloride, Tetraspasmin-Lefa |
| InChI Formula | InChI=1S/C24H31NO4.ClH/c1-5-26-21-10-9-17(14-22(21)27-6-2)13-20-19-16-24(29-8-4)23(28-7-3)15-18(19)11-12-25-20;/h9-10,13-16,25H,5-8,11-12H2,1-4H3;1H/b20-13-; |
| SMILES | Cl.CCOc1ccc(\C=C\2/NCCc3cc(OCC)c(OCC)cc23)cc1OCC |
| IUPAC Name | (1Z)-1-[(3,4-diethoxyphenyl)methylidene]-6,7-diethoxy-3,4-dihydro-2H-isoquinoline;hydrochloride |
| Molecular Weight (g/mol) | 433.97 |
| Show More |