Learn More
Drostanolone, TRC
Product Code. 30376787
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30376787 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30376787 Supplier Toronto Research Chemicals Supplier No. TRCD6895705MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Drostanolone
CAS 58-19-5
Molecular Formula C20 H32 O2
Quantity 5 mg
Synonym (2α,5α,17β)-17-Hydroxy-2-methylandrostan-3-one, 5α-Androstan-3-one, 17β-hydroxy-2α-methyl- (6CI,7CI,8CI), (2α,5α,17β)-17-Hydroxy-2-methylandrostan-3-one, 17β-Hydroxy-2α-methyl-5α-androstan-3-one, 2α-Methyl-17β-hydroxy-5α-androstan-3-one, 2α-Methyl-17β-hydroxyandrostan-3-one, 2α-Methyl-5α-androstan-17β-ol-3-one, 2α-Methyl-5α-dihydrotestosterone, Dihydro-2α-methyltestosterone, Dromostanolone, Drostanolone, Medrotestron, Metholone
InChI Formula InChI=1S/C20H32O2/c1-12-11-20(3)13(10-17(12)21)4-5-14-15-6-7-18(22)19(15,2)9-8-16(14)20/h12-16,18,22H,4-11H2,1-3H3/t12-,13+,14+,15+,16+,18+,19+,20+/m1/s1
SMILES C[C@@H]1C[C@@]2(C)[C@@H](CC[C@H]3[C@@H]4CC[C@H](O)[C@@]4(C)CC[C@H]23)CC1=O
IUPAC Name (2R,5S,8R,9S,10S,13S,14S,17S)-17-hydroxy-2,10,13-trimethyl-1,2,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydrocyclopenta[a]phenanthren-3-one
Molecular Weight (g/mol) 304.467
Formula Weight 304.24