missing translation for 'onlineSavingsMsg'
Learn More

DREADD agonist 21, MedChemExpress

Product Code. 16435607
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
Unit Size:
10mg
2mg
5mg
This item is not returnable. View return policy

Product Code. 16435607

Brand: Medchem Express HY100234

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress DREADD agonist 21 is a potent human muscarinic acetylcholine M3 receptors (hM3Dq) agonist (EC50=1.7 nM).

DREADD agonist 21 is a potent human muscarinic acetylcholine M3 receptors (hM3Dq) agonist (EC50=1.7 nM).

Specifications

Chemical Name or Material DREADD agonist 21
Physical Form Solid
CAS 56296-18-5
Color Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C17H18N4
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : ≥ 78 mg/mL (280.22 mM)
SMILES C12=CC=CC=C1NC3=CC=CC=C3C(N4CCNCC4)=N2
Molecular Weight (g/mol) 278.35
Formula Weight 278.35
Percent Purity 98.95%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.