Learn More
DRB1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA554209
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: VDSLDEPPNS RIFLVISKYT PESVLRERFS PFGDIQDIWV VRDKHTKESK GIAFVKFARS SQACRAMEEM HGQCLGPNDT KPIKVFIAQS RSSGSHRDVE DEELTRIFVM IPKSYTEE Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 98%.
DRB1 is RNA-binding protein with binding specificity for poly(C) and may play an important role in neural development.Specifications
DRB1 | |
Polyclonal | |
Unconjugated | |
RBM45 | |
developmentally regulated RNA binding protein 1; developmentally regulated RNA-binding protein 1; developmentally-regulated RNA-binding protein 1; dopamine receptor binding 1; Drb1; Drbp1; G430095G15Rik; putative RNA binding protein RB-1; RB-1; Rbm45; RNA binding motif protein 45; RNA-binding motif protein 45; RNA-binding protein 45 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
129831 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IUH3 | |
RBM45 | |
Recombinant protein corresponding to Human DRB1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only