Learn More
Julabo™ Drain Tap with Tube for Shaking Water Bath
Product Code. 10237373
Change view
Click to view available options
Type:
Drain tap with tube
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
10237373 Drain tap with tube Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10237373 Supplier Julabo™ Supplier No. 8970453
  • ONLINE EXCLUSIVE PRICE
    128.00 €
    112.00 € / Pack of 1
    Save 16.00 €
    12% Off
Estimated Shipment: 02-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
For Use With (Equipment) TW8, TW12, TW20 water baths, SW22, SW23 shaking water baths, HE-4, HL-4, MA-12, MA-26, MA-4 heating circulator
Type Drain tap with tube