missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DPP6 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17257903 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17257903 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17257903 Supplier Invitrogen™ Supplier No. PA5115140

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total DPP6 - Catalytic domain.

DPP6 encodes a single-pass type II membrane protein that is a member of the S9B family in clan SC of the serine proteases. This protein has no detectable protease activity, most likely due to the absence of the conserved serine residue normally present in the catalytic domain of serine proteases. However, it does bind specific voltage-gated potassium channels and alters their expression and biophysical properties. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DPP6
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene DPP6
Gene Accession No. P42658, P46101, Q9Z218
Gene Alias B930011P16Rik; D5Buc3; D5Buc4; D5Buc5; dipeptidyl aminopeptidase IV-related protein; dipeptidyl aminopeptidase-like protein 6; Dipeptidyl aminopeptidase-related protein; Dipeptidyl peptidase 6; Dipeptidyl peptidase IV-like protein; dipeptidyl peptidase IV-related protein; dipeptidyl peptidase like 6; dipeptidyl peptidase VI; dipeptidylpeptidase 6; dipeptidyl-peptidase 6; dipeptidylpeptidase VI; DPL1; DPP VI; Dpp6; Dpp-6; DPPX; Gm1377; In(5)6H p; In(5)6H-p; inversion, Chr 5, Harwell 6, proximal; MRD33; Peplb; Rw; VF2
Gene Symbols DPP6
Host Species Rabbit
Immunogen A synthesized peptide derived from human DPP6(Accession P42658), corresponding to amino acid residues Y205-I255.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13483, 1804, 29272
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.