missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DPP2 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA547489
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs and Western blots, approximately 60% cross-reactivity with recombinant mouse DPPII is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
DPP2 Polyclonal antibody specifically detects DPP2 in Human samples. It is validated for Immunoprecipitation, Western Blot.Specifications
DPP2 | |
Polyclonal | |
Unconjugated | |
DPP7 | |
carboxytripeptidase; dipeptidyl aminopeptidase II; dipeptidyl arylamidase II; dipeptidyl peptidase 2; Dipeptidyl peptidase 7; dipeptidyl peptidase II; dipeptidyl-peptidase 2; dipeptidylpeptidase 7; dipeptidyl-peptidase II; DPP II; DPP2; Dpp7; DPPII; QPP; Quiescent cell proline dipeptidase | |
Goat | |
Antigen affinity chromatography | |
RUO | |
29952 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q9UHL4 | |
DPP7 | |
Mouse myeloma cell line NS0-derived recombinant human DPPII/QPP/DPP7 Gly22-Leu492 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |