Learn More
DPH2 Monoclonal Antibody (OTI4E7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504852
Description
This gene is one of two human genes similar to the yeast gene dph2. The yeast gene was identified by its ability to complement a diphthamide mutant strain, and thus probably functions in diphthamide biosynthesis. Diphthamide is a post-translationally modified histidine residue present in elongation factor 2 (EF2) that is the target of diphtheria toxin ADP-ribosylation. Two transcript variants encoding different isoforms have been found for this gene.Specifications
| DPH2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q568Y2, Q9BQC3, Q9CR25 | |
| Dph2 | |
| Full length human recombit protein of human DPH2 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Canine, Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI4E7 | |
| Unconjugated | |
| DPH2 | |
| DPH2L2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 1802, 298452, 475387, 67728 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.