Learn More
Lovibond™ DPD 3 Solution
Click to view available options
Quantity:
15 mL
Unit Size:
15mL
Description
Lovibond DPD 3 Solution, Quantity: 15mL, Packaging: 1Each, Red
Specifications
Specifications
| Color | Red |
| Packaging | 1 Each |
| Quantity | 15 mL |