Learn More
DPC-681, MedChemExpress
Product Code. 16493727
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16493727 50 mg 50mg
16451809 1 mg 1mg
16440228 5 mg 5mg
16413646 25 mg 25mg
16407768 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16493727 Supplier Medchem Express Supplier No. HY19400
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress DPC-681 is a potent and selective inhibitor of HIV protease with IC90s for wild-type HIV-1 of 4 to 40 nM.
DPC-681 is a potent and selective inhibitor of HIV protease with IC90s for wild-type HIV-1 of 4 to 40 nM.
Specifications
Chemical Name or Material DPC-681
Physical Form Solid
CAS 284661-68-3
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C35H48FN5O5S
For Use With (Application) COVID-19-anti-virus
Synonym DPH-153893
Solubility Information DMSO : 100 mg/mL (149.29 mM; Need ultrasonic)
SMILES O=S(C1=CC(N)=CC=C1)(N(C[C@@H](O)[C@H](CC2=CC=CC=C2)NC([C@H](C(C)(C)C)NC(CNCC3=CC=CC(F)=C3)=O)=O)CC(C)C)=O
Molecular Weight (g/mol) 669.85
Formula Weight 669.85
Percent Purity 99.72%
Grade Research