Learn More
Doxybetasol, TRC
Product Code. 30383952
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30383952 50 mg
30379760 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30383952 Supplier Toronto Research Chemicals Supplier No. TRCD56100050MG
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1879-77-2
Molecular Formula C22 H29 F O4
Molecular Weight (g/mol) 376.46
Synonym (11β,16β)-9-Fluoro-11,17-dihydroxy-16-methyl-pregna-1,4-diene-3,20-dione, Pregna-1,4-diene-3,20-dione, 9-fluoro-11β,17-dihydroxy-16β-methyl- (7CI,8CI), 21-Deoxybetamethasone, 9-Fluoro-11β,17-dihydroxy-16β-methylpregna-1,4-diene-3,20-dione, Doxibetasol, Doxybetasol, Betamethasone Valerate Imp. B (EP)
IUPAC Name (8S,9R,10S,11S,13S,14S,16S,17R)-17-acetyl-9-fluoro-11,17-dihydroxy-10,13,16-trimethyl-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-3-one
SMILES C[C@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)C
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C22H29FO4/c1-12-9-17-16-6-5-14-10-15(25)7-8-19(14,3)21(16,23)18(26)11-20(17,4)22(12,27)13(2)24/h7-8,10,12,16-18,26-27H,5-6,9,11H2,1-4H3/t12-,16-,17-,18-,19-,20-,21-,22-/m0/s1
Formula Weight 376.205
Chemical Name or Material Doxybetasol