Description
This product has been discontinued. For the latest electronic dosimeter, see our EPD TruDose.
The high quality of the Thermo Scientific EPD Mk2, combined with the included easy-to-use Easy Issue and EasyEPD2 software packages and the integrated infrared EPD reader, makes the Dosimetry Solution the perfect kit for any facility looking to monitor radiation dose.
- One kit provides a total dosimetry solution
- Advanced radiological performance
- Proven technology Small and very lightweight dosimeter
- EPD operates on a single battery
- Rugged ergonomic design
- Integrated infrared reader in carry case
- EasyEPD and Easy Issue software included
Components:
Electronic Personal Dosimeter
- Mk2, G or N2 dosimeter models can be selected; standard kit contains 5 EPDs, with option to select up to 12 EPDs
- Multi-detector technology
- Excellent response to all radiation types; choice of beta/gamma, gamma/neutron, or gamma-only EPD
- Lightweight design: 95 g (3.2 oz) including battery and clip
- Size: 85 mm x 63 mm x 19 mm (3.3 x 2.5 x .08 in.), excluding clip
- Power supply: single AA battery 1.5V alkaline cell for up to 30 weeks of continuous operation OR 3.6 V lithium battery for typically 20 to 40 weeks continuous operation (interchangeable; life estimates depend on type)
- 10-year data retention without battery
- Configurable audible and visual alarms for dose, dose rate, count down time, read time, and failure modes
- Excellent immunity to electromagnetic interference
- Easy-to-read display with optional backlighting
EasyEPD2 Software
- No database required
- Configuration software to customize alarm levels, display information, and more
- Interfaces directly to the Thermo IrDA reader
- Automatic discovery and reading of a detected EPD unit
- Basic EPD Issue and Return functions supported
- Individual screen windows for each logical functional area of control
- Compatible with Microsoft™ Windows™
Easy Issue Software
- Flexible, simple and scalable database that can be stand-alone or networkable
- Supports grab-and-go operations
- Ability to allocate alarm levels for different tasks
- Restrict EPD issue based on personal radiation dose levels
EPD IrDA Reader
- Infrared EPD reader integrated into kit carrying case
- Reader software included
- Connect directly to a PC via serial interface
- Requires no power supply
Carrying Case
- Rugged Pelican case with molded foam inserts to protect EPD units
- Integrated EPD IrDA reader in case
- Easy storage for the entire system in one self-contained unit
- Can be shipped using FedEx or UPS
Ordering Information:
Building on the proven technology of the Thermo Scientific Mk2 Electronic Personal Dosimeter (EPD), the standard kit contains five EPDs, software to configure and issue the EPDs, and a rugged carrying case with an integrated IrDA reader. For larger facilities, there is an option to order from 5 to 12 Electronic Personal Dosimeters.
Includes:
Electronic Personal Dosimeter (Mk2, G or N2 dosimeter models can be selected; standard kit contains five EPDs, with option to select up to 12 EPDs); EasyEPD2 Software; Easy Issue Software; EPD IrDA Reader; Carrying Case
Recommended for:
- Medical walk-in clinics
- Radiology labs and stand-alone facilities
- Veteraniary offices
- Dental offices
- Ambulance companies
- First responders
- Research laboratories
Specifications
Specifications
| Product Type | Dosimetry Solution |