missing translation for 'onlineSavingsMsg'
Learn More

Dorsomorphin dihydrochloride, MedChemExpress

Product Code. 16406186
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16406186

Brand: Medchem Express HY13418

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Dorsomorphin dihydrochloride (BML-275 dihydrochloride; Compound C dihydrochloride) is a potent, selective and ATP-competitive AMPK inhibitor, with a Ki of 109 nM. Dorsomorphin dihydrochloride inhibits BMP pathway by targeting the type I receptors ALK2, ALK3, and ALK6. Dorsomorphin dihydrochloride induces autophagy.

Dorsomorphin dihydrochloride (BML-275 dihydrochloride

Specifications

Chemical Name or Material Dorsomorphin dihydrochloride
CAS 1219168-18-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C24H27Cl2N5O
Quantity 100 mg
Synonym Compound C dihydrochlorideBML-275 dihydrochloride
Solubility Information H2O : 50 mg/mL (105.84 mM; Need ultrasonic) ∣DMSO : 5 mg/mL (10.58 mM; Need ultrasonic)
SMILES [H]Cl.[H]Cl.C12=C(C3=CC=NC=C3)C=NN1C=C(C4=CC=C(OCCN5CCCCC5)C=C4)C=N2
Molecular Weight (g/mol) 472.41
Formula Weight 472.41
Percent Purity 98.76%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.