Learn More
Doramapimod, TRC
Specifications
Specifications
| Chemical Name or Material | Doramapimod |
| CAS | 285983-48-4 |
| Molecular Formula | C31H37N5O3 |
| Quantity | 10 mg |
| Synonym | N-[3-(1,1-Dimethylethyl)-1-(4-methylphenyl)-1H-pyrazol-5-yl]-N'-[4-[2-(4-morpholinyl)ethoxy]-1-naphthalenyl]urea, BIRB 796BS, Birb 796 |
| InChI Formula | InChI=1S/C31H37N5O3/c1-22-9-11-23(12-10-22)36-29(21-28(34-36)31(2,3)4)33-30(37)32-26-13-14-27(25-8-6-5-7-24(25)26)39-20-17-35-15-18-38-19-16-35/h5-14,21H,15-20H2,1-4H3,(H2,32,33,37) |
| SMILES | Cc1ccc(cc1)n2nc(cc2NC(=O)Nc3ccc(OCCN4CCOCC4)c5ccccc35)C(C)(C)C |
| Molecular Weight (g/mol) | 527.66 |