missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dopamine D3R/DRD3 Antibody (JE33-33), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32277
This item is not returnable.
View return policy
Description
Dopamine D3R/DRD3 Monoclonal antibody specifically detects Dopamine D3R/DRD3 in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| Dopamine D3R/DRD3 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000-1:2000 | |
| D(3) dopamine receptor, D3DR, Dopamine D3 receptor, dopamine receptor D3, essential tremor 1, ETM1, FET1, MGC149204, MGC149205 | |
| Synthetic peptide within Human Dopamine D3R/DRD3 aa 1-50/ 400. (Uniprot: P35462) | |
| 100 μL | |
| GPCR | |
| 1814 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| JE33-33 | |
| Unconjugated | |
| 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction