missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dop1R1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17920483
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17920483 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17920483 Supplier Invitrogen™ Supplier No. PA5144383

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Dop1R1 Polyclonal Antibody for Western Blot, ELISA

Receptor for dopamine. The activity of this receptor is mediated by G proteins which activate adenylyl cyclase. Might be involved in the processing of visual information and/or visual learning. Important for Pavlovian conditioning: required in the mushroom body as a receptor conveying unconditional stimuli information, has a role in memory formation for aversive and appetitive learning. Sleep-deprivation-induced impairments in learning can be partially explained through alterations in dopamine signaling, Dop1R1 expression levels are reduced; sleep may have a role in restoring dopamine homeostasis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dop1R1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 4.486 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene Dop1R1
Gene Accession No. P41596
Gene Alias DopR; DopR1; DopR35EF
Gene Symbols Dop1R1
Host Species Rabbit
Immunogen Recombinant Drosophila melanogaster (Fruit fly) Dop1R1 protein (20-142aa ).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 41726
Target Species Drosophila
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.