Learn More
BINDER™ Door Access System
Product Code. 16189780 Shop All BINDER Products
Click to view available options
:
For Use With: BINDER™ units series UFV
Product Code. 16189780
Brand: BINDER™ 80121899
Only null left
Add to basket
Add to basket
Door access system consisting of electromechanical door locking mechanism and electronic access control via NumPad
  • Consists of electromechanical door locking mechanism and electronic access control
  • Suitable for:
    • UF V 500
    • UF V 700
Specifications
Description Door access system - consisting of electromechanical door locking mechanism and electronic access control via NumPad
Type Door access system electr-mech UFV
For Use With (Equipment) BINDER Chambers Series UFV