missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Domvanalimab Humanized Recombinant Human Monoclonal Antibody

Human Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA554874

Product Code. 30131974

  • 394.00 € / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Compatible with Direct ELISA.

Domvanalimab is an anti-TIGIT monoclonal antibody that blocks the interaction between TIGIT and its ligands, enhancing T-cell and NK cell activity to promote anti-tumor immune responses.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Domvanalimab Humanized
Recombinant Monoclonal
Unconjugated
AB 154; GS-0154
Recombinant Human TIGIT.
100 μg
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ELISA
1 mg/mL
PBS with no preservative
Human
Protein A/G
RUO
Human
Antibody
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.