Learn More
Dolutegravir intermediate-1, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
Description
Dolutegravir intermediate-1 is a synthetic intermediate of Dolutegravir extracted from patent WO 2016125192 A2. Dolutegravir is an integrase inhibitor developed for the treatment of human immunodeficiency virus (HIV)-1 infection.
Specifications
Specifications
| Chemical Name or Material | Dolutegravir intermediate-1 |
| Physical Form | Solid |
| CAS | 1335210-23-5 |
| Color | White |
| Quantity | 5 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C13H17NO8 |
| Solubility Information | DMSO : 100 mg/mL (317.18 mM; Need ultrasonic) |
| SMILES | O=C(C1=C(OC)C(C(C(O)=O)=CN1CC(OC)OC)=O)OC |
| Molecular Weight (g/mol) | 315.28 |
| Show More |