missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DOK3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA521867
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: A431. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
DOK3 DOK proteins are enzymatically inert adaptor or scaffolding proteins. They provide a docking platform for the assembly of multimolecular signaling complexes. DOK3 is a negative regulator of JNK signaling in B-cells through interaction with INPP5D/SHIP1. May modulate ABL1 function.Specifications
DOK3 | |
Polyclonal | |
Unconjugated | |
DOK3 | |
AI450713; Docking protein 3; Dok3; Dokl; DOK-L; Dok-like protein; downstream of tyrosine kinase 3; FLJ22570; FLJ39939; p62(dok)-like protein; p62Dok-like protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79930 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q7L591 | |
DOK3 | |
Synthetic peptide corresponding to a region within amino acids 434 and 496 of DOK3 (Uniprot ID#Q7L591) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |