missing translation for 'onlineSavingsMsg'
Learn More

DOK3 Monoclonal Antibody (OTI1H9), OriGene

Product Code. 17507854
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
Unit Size:
30µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17507854 30 μL 30µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17507854 Supplier Origene Technologies Supplier No. TA813133S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

DOK3 Monoclonal antibody specifically detects DOK3 in Human samples. It is validated for Western Blot

Specifications

Antigen DOK3
Applications Western Blot
Classification Monoclonal
Clone OTI1H9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene CD86
Gene Accession No. P42081
Gene Alias AI450713; Docking protein 3; Dok3; Dokl; DOK-L; Dok-like protein; downstream of tyrosine kinase 3; FLJ22570; FLJ39939; p62(dok)-like protein; p62Dok-like protein
Host Species Mouse
Immunogen Full length human recombinant protein of human CD86 (NP_787058) produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 942
Target Species Human
Content And Storage -20°C
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.