missing translation for 'onlineSavingsMsg'
Learn More

Dofequidar fumarate, MedChemExpress

Product Code. p-200096842
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16497226 100 mg 100mg
16449787 5 mg 5mg
16447268 10 mg 10mg
16403287 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16497226 Supplier Medchem Express Supplier No. HY17013A
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Dofequidar fumarate(MS-209 fumarate), an orally active quinoline compound, has been reported to overcome MDR by inhibiting ABCB1/P-gp, ABCC1/MDR-associated protein 1, or both.

Dofequidar fumarate(MS-209 fumarate), an orally active quinoline compound, has been reported to overcome MDR by inhibiting ABCB1/P-gp, ABCC1/MDR-associated protein 1, or both.

Specifications

Chemical Name or Material Dofequidar fumarate
CAS 153653-30-6
Color White
Purity Grade Notes Research
Molecular Formula C34H35N3O7
Quantity 100 mg
Synonym MS-209
Solubility Information DMSO : 100 mg/mL (167.32 mM; Need ultrasonic) ∣H2O : 1 mg/mL (1.67 mM; Need ultrasonic)
SMILES OC(COC1=C2C=CC=NC2=CC=C1)CN3CCN(C(C(C4=CC=CC=C4)C5=CC=CC=C5)=O)CC3.O=C(O)/C=C/C(O)=O
Molecular Weight (g/mol) 597.66
Formula Weight 597.66
Percent Purity 98.4%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.