Learn More
Dodecylbenzenesulfonic Acid (mixture of isomers) (Technical Grade), TRC
Product Code. 30135993
Change view
Click to view available options
Quantity:
50 mL
100 mL
250 mL
Unit Size:
100mL
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135993 250 mL 250mL
30137806 100 mL 100mL
30008732 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30135993 Supplier Toronto Research Chemicals Supplier No. TRCD494568250ML
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 121-65-3
Molecular Formula C18H30O3S
Molecular Weight (g/mol) 326.49
Synonym 4-Dodecylbenzenesulfonic Acid, Benzenesulfonic acid, 4-dodecyl-, Benzenesulfonic acid, p-dodecyl- (8CI), 4-Dodecylbenzenesulfonic acid, NSC 85535, Naxcat 1270, p-Dodecylbenzenesulfonic acid
IUPAC Name 4-dodecylbenzenesulfonic acid
SMILES CCCCCCCCCCCCc1ccc(cc1)S(=O)(=O)O
Specifications
Quantity 250 mL
InChI Formula InChI=1S/C18H30O3S/c1-2-3-4-5-6-7-8-9-10-11-12-17-13-15-18(16-14-17)22(19,20)21/h13-16H,2-12H2,1H3,(H,19,20,21)
Formula Weight 326.1916
Chemical Name or Material Dodecylbenzenesulfonic Acid (mixture of isomers) (Technical Grade)