Learn More
Dodecylbenzene-d5, TRC
Specifications
Specifications
| Chemical Name or Material | Dodecylbenzene-d5 |
| CAS | 2733776-98-0 |
| Molecular Formula | C18H25D5 |
| Quantity | 25 mg |
| Synonym | 1-Dodecylbenzene-d5, 1-Phenyl-n-dodecane-d5, 1-Phenyldodecane-d5, Alkylate P 1-d5, Detergent Alkylate No. 2-d5, Laurylbenzene-d5, Nalkylene 500-d5, n-Dodecylbenzene-d5 |
| InChI Formula | InChI=1S/C18H30/c1-2-3-4-5-6-7-8-9-10-12-15-18-16-13-11-14-17-18/h11,13-14,16-17H,2-10,12,15H2,1H3/i11D,13D,14D,16D,17D |
| SMILES | CCCCCCCCCCCCC1=C([2H])C([2H])=C([2H])C([2H])=C1[2H] |
| IUPAC Name | 1-dodecylbenzene-2,3,4,5,6-d5 |
| Molecular Weight (g/mol) | 251.46 |
| Formula Weight | 251.27 |