| InChI Formula |
InChI=1S/C22H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22(23)24/h3-4,6-7,9-10,12-13,15-16,18-19H,2,5,8,11,14,17,20-21H2,1H3,(H,23,24)/b4-3-,7-6-,10-9-,13-12-,16-15-,19-18- |
| Synonym |
4,7,10,13,16,19-Docosahexaenoic acid, (4Z,7Z,10Z,13Z,16Z,19Z)-, 4,7,10,13,16,19-Docosahexaenoic acid, (all-Z)- (8CI), Docosahexaenoic acid (6CI), (4Z,7Z,10Z,13Z,16Z,19Z)-4,7,10,13,16,19-Docosahexaenoic acid, (4Z,7Z,10Z,13Z,16Z,19Z)-Docosahexaenoic acid, (4Z,7Z,10Z,13Z,16Z,19Z)-Docosahexenoic acid, (all-Z)-4,7,10,13,16,19-Docosahexaenoic acid, 4-cis,7-cis,10-cis,13-cis,16-cis,19-cis-Docosahexaenoic acid, AlgaMac 3050, Algatrium, AquaGrow Advantage, C22:6, Cervonic acid, DHA, DHA 22, DHA 22HG, DHA 46, DHA 46A, DHA S Rosemary Sun, DHASCO, DHASCO-HM, DHASCO-S, DHASCO-T, Doconexent, Efalex, Marinol D 50TG, Martek DHA HM, Martek DHA Powder KS 35, Martek DHA Powder KSF 58, Martek DHA-S, Maxigard, Monolife 50, Monolife 90, Ropufa 60, S.Presso, all-Z-Docosahexaenoic acid, all-cis-4,7,10,13,16,19-Docosahexaenoic acid, δ4,7,10,13,16,19-Docosahexaenoic acid |