missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Docaravimab Recombinant Mouse Monoclonal Antibody
Mouse Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA554873
This item is not returnable.
View return policy
Description
Compatible with Direct ELISA.
Docaravimab is a monoclonal antibody that neutralizes SARS-CoV-2 by binding to the spike protein, preventing the virus from entering human cells.
Specifications
| Docaravimab | |
| Recombinant Monoclonal | |
| Unconjugated | |
| ERA glycoprotein ectodomain epitope G-III; Evelyn-Rockitniki-Abelseth | |
| Recombinant RABV G/Glycoprotein. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA | |
| 3.15 mg/mL | |
| PBS with no preservative | |
| Mouse | |
| Protein A/G | |
| RUO | |
| Virus | |
| Antibody | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction